Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023049.1 | NP_075536.1 | 10 | 2652 | ||
GenBank mRNA | AK003566.1 | - | 3 | 852 | ||
AK083400.1 | - | 8 | 2176 | |||
AK155235.1 | - | 6 | 2366 | |||
BC031161.1 | - | 10 | 2652 | 4981777 |