Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 7 | 1274 | ||
NM_001164208.1 | NP_001157680.1 | 7 | 922 | |||
NM_001164209.2 | NP_001157681.1 | 8 | 1130 | |||
NM_001286651.1 | NP_001273580.1 | 8 | 1127 | |||
NM_001286652.1 | NP_001273581.1 | 7 | 1034 | |||
NM_023056.4 | NP_075543.1 | 7 | 1523 | |||
GenBank mRNA | AB031386.1 | - | 7 | 1083 | ||
AK007408.1 | - | 7 | 929 | |||
AK156042.1 | - | 7 | 1275 | |||
BC019539.1 | - | 6 | 892 | 4237639 | ||
BC022166.1 | - | 7 | 1538 | 3582123 |