Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lima1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113545.1 | NP_001107017.1 | 11 | 3348 | ||
NM_023063.4 | NP_075550.2 | 8 | 3220 | |||
GenBank mRNA | AF307844.1 | - | 8 | 3142 | ||
AK132314.1 | - | 8 | 3166 | |||
AK167220.1 | - | 8 | 2779 | |||
AK194816.1 | - | 1 | 85 |