Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023290.2 | NP_075779.2 | 8 | 1259 | ||
GenBank mRNA | AF277171.1 | - | 8 | 1227 | ||
AK009515.1 | - | 8 | 1267 | |||
AK011295.1 | - | 8 | 1238 | |||
AK077926.1 | - | 8 | 1259 | |||
BC025547.1 | - | 8 | 1278 | 5320887 |