Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retnlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023881.4 | NP_076370.3 | 2 | 315 | ||
GenBank mRNA | AF290871.1 | - | 2 | 317 | ||
AF323083.1 | - | 2 | 202 | |||
AK020240.1 | - | 2 | 203 | |||
BC022650.1 | - | 2 | 185 | 4216606 | ||
DQ157777.1 | - | 1 | 74 |