Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006682.2 | NP_006673.1 | 1 | 156 | ||
GenBank mRNA | AK222999.1 | - | 1 | 124 | ||
AK310921.1 | - | 1 | 171 | |||
AK312672.1 | - | 1 | 128 | |||
BC017813.1 | - | 1 | 140 | |||
BC033820.1 | - | 1 | 144 | 5219649 | ||
BC073986.1 | - | 1 | 129 | 5748616 | ||
DB247183.1 | - | 1 | 171 | |||
Z36531.1 | - | 1 | 149 |