Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 0610009O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024179.5 | NP_077141.2 | 13 | 2361 | ||
GenBank mRNA | AK002437.1 | - | 13 | 2351 | ||
AK009400.1 | - | 6 | 1373 | |||
AK171837.1 | - | 13 | 2352 | |||
AK172900.1 | - | 13 | 2286 | |||
AU257559.1 | - | 1 | 78 |