Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025359.3 | NP_079635.1 | 6 | 1582 | ||
GenBank mRNA | AK008175.1 | - | 6 | 1522 | ||
AK090217.1 | - | 6 | 1594 | |||
AK090322.1 | - | 6 | 1594 | |||
AK165768.1 | - | 6 | 1618 | |||
AK167495.1 | - | 6 | 1634 | |||
AK167979.1 | - | 6 | 1585 | |||
BC018317.1 | - | 6 | 1581 | 4166625 |