Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303471.2 | NP_001290400.1 | 3, 4 | 614 | ||
NM_001305557.1 | NP_001292486.1 | 2, 3 | 441 | |||
NM_001305563.1 | NP_001292492.1 | 3, 4 | 254 | |||
NM_001305564.1 | NP_001292493.1 | 2, 3 | 197 | |||
NM_006701.4 | NP_006692.1 | 2, 3 | 465 | |||
GenBank mRNA | AF023611.1 | - | 2, 3 | 391 | ||
AF146373.1 | - | 2, 3 | 265 | |||
AK308703.1 | - | 2, 3 | 177 | |||
AK309098.1 | - | 3, 4 | 531 | |||
AK314901.1 | - | 2, 3 | 417 | |||
AV729855.2 | - | 2, 3 | 197 | |||
BC001046.2 | - | 2, 3 | 405 | 2823293 | ||
BC019272.1 | - | 2, 3 | 405 | 2823293 | ||
BE385528.1 | - | 2, 3 | 401 | |||
BF972457.1 | - | 2, 3 | 466 | |||
BM922767.1 | - | 2, 3 | 442 |