Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Anapc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038230.2 | NP_001033319.1 | 3 | 2723 | ||
NM_025389.4 | NP_079665.1 | 3 | 2568 | |||
GenBank mRNA | AK045408.1 | - | 3 | 2511 |