Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025480.3 | NP_079756.2 | 5 | 953 | ||
GenBank mRNA | AK012789.2 | - | 5 | 954 | ||
AK080853.1 | - | 5 | 954 | |||
AK162257.1 | - | 5 | 948 | |||
BC060969.1 | - | 5 | 942 | 6704414 | ||
BC110310.1 | - | 5 | 936 | 5325405 |