Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc90b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025515.3 | NP_079791.2 | 3, 4 | 711 | ||
GenBank mRNA | AK009370.1 | - | 3, 4 | 332 | ||
AK011631.1 | - | 2, 3 | 314 | |||
AK084673.1 | - | 3, 4 | 346 | |||
AK160230.1 | - | 3, 4 | 367 | |||
BC115973.1 | - | 3, 4 | 332 | 40100643 |