Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282601.1 | NP_001269530.1 | 4 | 655 | ||
NM_001282602.1 | NP_001269531.1 | 4 | 655 | |||
NM_001282604.1 | NP_001269533.1 | 5 | 704 | |||
NM_001282605.1 | NP_001269534.1 | 4 | 655 | |||
NM_006601.6 | NP_006592.3 | 4 | 655 | |||
NR_104219.1 | 4 | 992 | ||||
XM_005268576.4 | XP_005268633.1 | 4 | 2245 | |||
XM_006719199.2 | XP_006719262.1 | 4 | 1004 | |||
XM_011537773.2 | XP_011536075.1 | 4 | 563 | |||
XM_011537774.2 | XP_011536076.1 | 4 | 1004 | |||
XM_017018716.1 | XP_016874205.1 | 4 | 1004 | |||
GenBank mRNA | AK098214.1 | - | 4 | 992 | ||
AK291945.1 | - | 4 | 409 | |||
AK293139.1 | - | 3 | 514 | |||
AK298147.1 | - | 4 | 531 | |||
AK298160.1 | - | 4 | 469 | |||
AL603017.1 | - | 4 | 541 | |||
BC003005.1 | - | 4 | 352 | 2821965 | ||
BM541503.1 | - | 5 | 521 | |||
BU167210.1 | - | 4 | 510 | |||
L24804.1 | - | 4 | 478 |