Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025597.2 | NP_079873.1 | 3 | 285 | ||
GenBank mRNA | AK007930.1 | - | 3 | 284 | ||
AK009008.1 | - | 3 | 247 | |||
AK012320.1 | - | 3 | 286 | |||
AK013608.1 | - | 3 | 271 | |||
AK138123.1 | - | 3 | 283 | |||
BC028669.1 | - | 3 | 441 | |||
BC036989.1 | - | 3 | 282 | 4989233 |