Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aggf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025630.3 | NP_079906.2 | 4 | 942 | ||
GenBank mRNA | AK008168.1 | - | 4 | 915 | ||
AK008174.1 | - | 4 | 912 | |||
AK008399.1 | - | 4 | 915 | |||
AK017248.1 | - | 4 | 898 | |||
AK132575.1 | - | 4 | 943 | |||
AY500995.1 | - | 4 | 915 | |||
BC052410.1 | - | 4 | 919 | 5716440 |