Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025656.4 | NP_079932.2 | 8 | 746 | ||
GenBank mRNA | AK005928.1 | - | 8 | 748 | ||
AK007515.1 | - | 8 | 747 | |||
AK013414.1 | - | 8 | 703 | |||
BC053424.1 | - | 8 | 1175 | 6336522 |