Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025671.2 | NP_079947.1 | 7 | 1377 | ||
GenBank mRNA | AK004943.1 | - | 7 | 1378 | ||
AK018534.1 | - | 7 | 1378 | |||
AK168946.1 | - | 7 | 1295 | |||
BC022762.1 | - | 6 | 1225 |