Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spryd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025697.3 | NP_079973.1 | 5 | 922 | ||
GenBank mRNA | AK018044.1 | - | 5 | 799 | ||
AK018153.1 | - | 5 | 923 | |||
AK018290.1 | - | 5 | 772 | |||
AK137685.1 | - | 771 | ||||
AK150789.1 | - | 4 | 694 | |||
AK167306.1 | - | 5 | 764 | |||
AK169059.1 | - | 5 | 771 | |||
BC037015.1 | - | 5 | 756 | 5344409 |