Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spryd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025697.3 | NP_079973.1 | 5 | 1202 | ||
GenBank mRNA | AK018044.1 | - | 5 | 1078 | ||
AK018153.1 | - | 5 | 1203 | |||
AK018290.1 | - | 5 | 1052 | |||
AK137685.1 | - | 1051 | ||||
AK150789.1 | - | 4 | 974 | |||
AK167306.1 | - | 5 | 1044 |