Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacrgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025755.3 | NP_080031.1 | 6 | 540 | ||
GenBank mRNA | AK014276.1 | - | 6 | 553 | ||
AK016966.1 | - | 6 | 589 | |||
AK018101.1 | - | 6 | 549 | |||
AK032498.1 | - | 5 | 2288 | |||
BC018481.1 | - | 6 | 540 | 3482169 |