Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hint3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025798.3 | NP_080074.1 | 2 | 209 | ||
GenBank mRNA | AK002482.1 | - | 2 | 304 | ||
AK007758.1 | - | 2 | 226 | |||
AK027974.1 | - | 2 | 210 | |||
AY040768.1 | - | 2 | 212 | |||
BC025065.1 | - | 2 | 209 | 4035971 |