Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025811.3 | NP_080087.1 | 3 | 859 | ||
GenBank mRNA | AK002650.1 | - | 3 | 444 | ||
AK004564.1 | - | 3 | 506 | |||
AK033384.1 | - | 3 | 860 | |||
BC095956.1 | - | 3 | 803 | 5330002 | ||
BC138190.1 | - | 3 | 696 | 8861209 | ||
BC138191.1 | - | 3 | 696 | 8861210 |