Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025811.3 | NP_080087.1 | 4, 5 | 1327 | ||
GenBank mRNA | AK002650.1 | - | 4, 5 | 912 | ||
AK033384.1 | - | 4, 5 | 1328 | |||
BC041104.1 | - | 2, 3 | 296 | |||
BC050211.1 | - | 2, 3 | 183 | |||
BC095956.1 | - | 4, 5 | 1271 | 5330002 | ||
BC138190.1 | - | 4, 5 | 1164 | 8861209 | ||
BC138191.1 | - | 4, 5 | 1164 | 8861210 |