Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF373710.1 | - | 7 | 468 | ||
AK003572.2 | - | 7 | 498 | |||
AK003699.1 | - | 7 | 422 | |||
AK013852.1 | - | 7 | 419 | |||
AK085669.1 | - | 6 | 1138 | |||
BC013535.1 | - | 7 | 431 | 4239189 |