Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144826.1 | NP_001138298.1 | 11 | 1510 | ||
NM_006695.4 | NP_006686.1 | 11 | 1525 | |||
XM_017024038.1 | XP_016879527.1 | 12 | 1612 | |||
XM_017024039.1 | XP_016879528.1 | 12 | 1609 | |||
XM_017024040.1 | XP_016879529.1 | 11 | 1446 | |||
GenBank mRNA | AF055026.1 | - | 11 | 1289 | ||
AK226045.1 | - | 11 | 1486 | |||
BC006194.2 | - | 11 | 1488 | 2823366 | ||
BX537443.1 | - | 11 | 1407 | |||
U93871.1 | - | 11 | 1236 |