Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303471.2 | NP_001290400.1 | 4 | 687 | ||
NM_001305557.1 | NP_001292486.1 | 3 | 514 | |||
NM_001305563.1 | NP_001292492.1 | 4 | 327 | |||
NM_001305564.1 | NP_001292493.1 | 3 | 270 | |||
NM_006701.4 | NP_006692.1 | 3 | 538 | |||
NR_131175.1 | 5 | 757 | ||||
NR_131176.1 | 4 | 608 | ||||
NR_131177.1 | 2 | 434 | ||||
GenBank mRNA | AF023611.1 | - | 3 | 464 | ||
AF023612.1 | - | 4 | 534 | |||
AF146373.1 | - | 3 | 338 | |||
AK308703.1 | - | 3 | 250 | |||
AK309098.1 | - | 4 | 604 | |||
AK314901.1 | - | 3 | 490 | |||
AL522879.3 | - | 5 | 746 | |||
AV729855.2 | - | 3 | 270 | |||
AW796546.1 | - | 2 | 168 | |||
BC001046.2 | - | 3 | 478 | 2823293 | ||
BC019272.1 | - | 3 | 478 | 2823293 | ||
BE385528.1 | - | 3 | 474 | |||
BF972457.1 | - | 3 | 539 | |||
BM922767.1 | - | 3 | 515 | |||
BU553165.1 | - | 553 |