Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277273.1 | NP_001264202.1 | 5, 6 | 497 | ||
NM_025923.3 | NP_080199.1 | 5, 6 | 497 | |||
GenBank mRNA | AA476004.1 | - | 3, 4 | 168 | ||
AF513619.1 | - | 5, 6 | 497 | |||
AK008605.1 | - | 5, 6 | 383 | |||
AK011520.1 | - | 5, 6 | 402 | |||
AK011752.1 | - | 5, 6 | 374 | |||
AK011903.1 | - | 5, 6 | 380 | |||
AK017431.1 | - | 5, 6 | 380 | |||
AK018018.1 | - | 5, 6 | 379 | |||
AK076017.1 | - | 5, 6 | 380 | |||
AK168538.1 | - | 5, 6 | 381 | |||
BC032876.1 | - | 5, 6 | 371 | 1282316 | ||
BC066181.1 | - | 5, 6 | 426 | 6435171 | ||
CN455644.1 | - | 4, 5 | 283 |