Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277273.1 | NP_001264202.1 | 6, 7 | 604 | ||
NM_025923.3 | NP_080199.1 | 6, 7 | 604 | |||
GenBank mRNA | AA476004.1 | - | 4, 5 | 275 | ||
AF513619.1 | - | 6, 7 | 604 | |||
AK008605.1 | - | 6, 7 | 490 | |||
AK011520.1 | - | 6, 7 | 509 | |||
AK011752.1 | - | 6, 7 | 481 | |||
AK011903.1 | - | 6, 7 | 487 | |||
AK017431.1 | - | 6, 7 | 487 | |||
AK018018.1 | - | 6, 7 | 486 | |||
AK020981.1 | - | 2, 3 | 209 | |||
AK076017.1 | - | 6, 7 | 487 | |||
AK168538.1 | - | 6, 7 | 488 | |||
BC032876.1 | - | 6, 7 | 478 | 1282316 | ||
BC066181.1 | - | 6, 7 | 533 | 6435171 | ||
CN455644.1 | - | 5, 6 | 390 |