Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA472267.1 | - | 1 | 396 | ||
AK012312.1 | - | 7 | 1085 | |||
AK078139.1 | - | 7 | 1359 | |||
BC031761.1 | - | 7 | 1078 | 4235549 |