Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012312.1 | - | 7 | 1870 | ||
AK021210.1 | - | 1 | 436 | |||
AK046873.1 | - | 1 | 337 | |||
AK078139.1 | - | 7 | 2144 | |||
BC031761.1 | - | 7 | 1863 | 4235549 |