Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sclt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081411.1 | NP_001074880.1 | 3 | 621 | ||
GenBank mRNA | AK011907.1 | - | 3 | 419 | ||
AK014777.1 | - | 3 | 445 | |||
AK016469.1 | - | 3 | 619 | |||
AK083894.1 | - | 3 | 604 | |||
BC043136.1 | - | 3 | 588 | 6812105 | ||
BC060271.1 | - | 3 | 588 | 6812105 |