Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam229b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183254.1 | NP_899077.1 | 4, 5 | 505 | ||
GenBank mRNA | AK006353.1 | - | 4, 5 | 505 | ||
AK006431.1 | - | 4, 5 | 484 | |||
BC147171.1 | - | 3, 4 | 434 | 9056241 | ||
BC147172.1 | - | 3, 4 | 434 | 9056208 | ||
BC172004.1 | - | 3, 4 | 431 | 9054290 |