Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam229b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183254.1 | NP_899077.1 | 5 | 668 | ||
GenBank mRNA | AK006353.1 | - | 5 | 668 | ||
AK006431.1 | - | 5 | 647 |