Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ndrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145959.1 | NP_001139431.1 | 15 | 1416 | ||
NM_013864.2 | NP_038892.1 | 16 | 1458 | |||
GenBank mRNA | AA014560.1 | - | 1 | 59 | ||
AB033921.1 | - | 16 | 1321 | |||
AK003364.1 | - | 2 | 363 | |||
AK076514.1 | - | 13 | 1092 | |||
AK154597.1 | - | 16 | 1326 | |||
AK158900.1 | - | 15 | 1325 | |||
AK168011.1 | - | 15 | 1283 | |||
AK173136.1 | - | 13 | 1099 | |||
BC012963.1 | - | 16 | 1325 | 4211421 |