Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013867.2 | NP_038895.1 | 10, 11 | 2488 | ||
GenBank mRNA | AF179566.1 | - | 10, 11 | 2436 | ||
AK143894.1 | - | 10, 11 | 2276 | |||
AK158241.1 | - | 10, 11 | 2489 | |||
AK167578.1 | - | 10, 11 | 2306 | |||
BC023930.1 | - | 10, 11 | 2477 | 5324365 |