Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006636.3 | NP_006627.2 | 8 | 1197 | ||
NR_027405.1 | 9 | 1295 | ||||
XM_006711924.2 | XP_006711987.1 | 7 | 984 | |||
GenBank mRNA | AK130664.1 | - | 8 | 1145 | ||
AK223041.1 | - | 8 | 1145 | |||
AK300035.1 | - | 7 | 962 | |||
AK302245.1 | - | 6 | 837 | |||
BC001548.1 | - | 8 | 1132 | 3462450 | ||
BC015062.1 | - | 9 | 1245 | 4285669 | ||
BC017054.1 | - | 8 | 1125 | 3846710 | ||
X16396.1 | - | 8 | 1115 |