Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159630.1 | NP_001153102.1 | 5, 6 | 537 | ||
NM_013883.2 | NP_038911.1 | 5, 6 | 537 | |||
GenBank mRNA | AB030906.1 | - | 5, 6 | 436 | ||
AB221680.1 | - | 1, 2 | 78 | |||
AB221681.1 | - | 1, 2 | 78 | |||
BC034667.1 | - | 3, 4 | 247 | 5356322 |