Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013900.2 | NP_038928.1 | 16 | 2503 | ||
GenBank mRNA | AB024336.1 | - | 16 | 2503 | ||
AK036444.1 | - | 16 | 2438 | |||
BC040347.1 | - | 16 | 2426 | 3672916 |