Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012330.1 | NP_001012330.1 | 2 | 3611 | ||
NM_013915.3 | NP_038943.3 | 1 | 4717 | |||
GenBank mRNA | AF140224.1 | - | 1 | 4719 | ||
AK088899.1 | - | 1 | 2157 | |||
AK147200.1 | - | 2 | 3566 | |||
BC050909.1 | - | 2 | 3638 | |||
BC054529.1 | - | 1 | 3719 | 5721393 | ||
BC054742.1 | - | 1 | 3719 | 5721393 | ||
BC089333.1 | - | 3 | 3507 |