Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163447.1 | NP_001156919.1 | 11, 12 | 1418 | ||
NM_001163448.1 | NP_001156920.1 | 10, 11 | 1403 | |||
NM_001163449.1 | NP_001156921.1 | 12, 13 | 1352 | |||
NM_001163450.1 | NP_001156922.1 | 11, 12 | 1328 | |||
NM_001163451.1 | NP_001156923.1 | 11, 12 | 1325 | |||
NM_001163453.1 | NP_001156925.1 | 10, 11 | 1310 | |||
NM_013931.4 | NP_038959.2 | 11, 12 | 1421 | |||
GenBank mRNA | AB005662.1 | - | 11, 12 | 1308 | ||
AB043123.1 | - | 12, 13 | 1229 | |||
AB043124.1 | - | 11, 12 | 1298 | |||
AB043125.1 | - | 11, 12 | 1295 | |||
AB093282.2 | - | 11, 12 | 1280 | |||
AF178636.1 | - | 10, 11 | 1320 | |||
AF178637.1 | - | 11, 12 | 1414 | |||
AF262046.1 | - | 11, 12 | 1404 | |||
BC060603.1 | - | 12, 13 | 1287 | 5706805 | ||
BC065105.1 | - | 10, 11 | 1300 | 30356817 | ||
BC068312.1 | - | 11, 12 | 1399 | 30100914 |