Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015773.2 | NP_056588.1 | 5 | 780 | ||
GenBank mRNA | AF173866.1 | - | 5 | 717 | ||
AK005732.1 | - | 5 | 728 | |||
AK054199.1 | - | 4 | 816 | |||
BC090660.1 | - | 5 | 723 | 30673350 | ||
BC100455.1 | - | 5 | 730 | 30919921 |
Gene Symbol: Spag6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001334.2 | NP_001001334.2 | 5 | 684 | ||
GenBank mRNA | AK132882.1 | - | 5 | 684 | ||
BC061194.1 | - | 5 | 683 |