Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015773.2 | NP_056588.1 | 11 | 2105 | ||
GenBank mRNA | AF173866.1 | - | 11 | 2042 | ||
BC090660.1 | - | 11 | 2048 | 30673350 | ||
BC100455.1 | - | 11 | 2055 | 30919921 |