Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs6st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077202.2 | NP_001070670.1 | 5 | 1553 | ||
NM_001290467.1 | NP_001277396.1 | 3 | 1433 | |||
NM_001290468.1 | NP_001277397.1 | 4 | 1194 | |||
NM_015819.4 | NP_056634.3 | 2 | 1074 | |||
GenBank mRNA | AB024565.1 | - | 4 | 1116 | ||
AK043774.1 | - | 4 | 677 | |||
AK083281.1 | - | 4 | 1182 | |||
AK171680.1 | - | 2 | 1077 | |||
BC037659.1 | - | 2 | 1025 | |||
BC047151.1 | - | 3 | 1203 | |||
BC063327.1 | - | 2 | 1066 | 6531439 |