Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015821.2 | NP_056636.2 | 3 | 1507 | ||
GenBank mRNA | AF176523.1 | - | 3 | 1432 | ||
AK053206.1 | - | 4 | 1675 | |||
AK155194.1 | - | 3 | 1468 | |||
AK170367.1 | - | 3 | 1467 | |||
BC023902.1 | - | 2 | 1372 | 5321302 |