Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016699.2 | NP_057908.2 | 5 | 547 | ||
GenBank mRNA | AF091392.1 | - | 5 | 516 | ||
AK017621.1 | - | 5 | 520 | |||
AK081014.1 | - | 5 | 543 | |||
AK144162.1 | - | 5 | 532 | |||
AK162887.1 | - | 5 | 516 | |||
AK166070.1 | - | 5 | 531 | |||
AK168700.1 | - | 5 | 520 | |||
AK207676.1 | - | 3 | 153 | |||
BC027326.1 | - | 5 | 525 | 3987812 | ||
BC027840.1 | - | 5 | 535 | 5325071 |