Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038710.1 | NP_001033799.1 | 8 | 1205 | ||
NM_016711.3 | NP_057920.2 | 8 | 1141 | |||
GenBank mRNA | AF177170.1 | - | 8 | 1090 | ||
AF237629.1 | - | 9 | 1049 | |||
AK015263.1 | - | 8 | 1185 | |||
AK017072.1 | - | 8 | 1189 | |||
AK076562.1 | - | 8 | 1155 | |||
AK079077.1 | - | 8 | 985 | |||
AK147581.1 | - | 8 | 1137 | |||
AK164178.1 | - | 8 | 987 | |||
BC052227.1 | - | 8 | 1205 | |||
BC061124.1 | - | 8 | 971 | 6531518 |