Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016847.2 | NP_058543.2 | 2 | 2207 | ||
GenBank mRNA | AK137203.1 | - | 2 | 2135 | ||
AK149578.1 | - | 2 | 2150 | |||
AK155952.1 | - | 2 | 2139 | |||
BC024149.1 | - | 2 | 2205 | 4220075 | ||
D49730.1 | - | 2 | 2220 |
Set Membership: |
G-protein coupled receptor |