Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016963.2 | NP_058659.1 | 9 | 1206 | ||
GenBank mRNA | AF177172.1 | - | 9 | 1194 | ||
AF237630.1 | - | 9 | 1120 | |||
AK017725.1 | - | 9 | 1202 | |||
AK050372.1 | - | 9 | 1208 | |||
AK084463.1 | - | 6 | 728 | |||
AK147017.1 | - | 9 | 1207 | |||
AK167723.1 | - | 9 | 1204 | |||
AK169218.1 | - | 9 | 1207 | |||
AK187754.1 | - | 3 | 304 | |||
BC082595.1 | - | 9 | 1198 | 30645500 |