Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113486.1 | NP_001106958.1 | 10 | 1605 | ||
NM_001113487.1 | NP_001106959.1 | 9 | 1785 | |||
NM_001113488.1 | NP_001106960.1 | 8 | 954 | |||
NM_017380.2 | NP_059076.1 | 8 | 935 | |||
GenBank mRNA | AJ250723.1 | - | 8 | 941 | ||
AK031757.1 | - | 8 | 830 | |||
AK122415.2 | - | 9 | 1579 | |||
AK141312.1 | - | 10 | 1546 | |||
AK164581.1 | - | 8 | 840 | |||
AK211632.1 | - | 47 | ||||
BC046524.1 | - | 10 | 1605 | 5290621 | ||
BC094455.1 | - | 11 | 1309 |