Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018810.2 | NP_061280.2 | 2 | 380 | ||
GenBank mRNA | AB026986.1 | - | 2 | 485 | ||
AF192785.1 | - | 2 | 405 | |||
AK005137.1 | - | 2 | 214 | |||
AK029500.1 | - | 2 | 420 | |||
AK077060.1 | - | 2 | 381 | |||
AK078962.1 | - | 2 | 310 | |||
BC003329.1 | - | 2 | 369 | 3158135 |